Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scpl42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_123589.4 | NP_199039.1 | 10 - 11 | 1933 | 92 | |
GenBank mRNA | AF361604.1 | - | 10 - 11 | 1326 | 92 | |
AY143899.1 | - | 9 - 10 | 1255 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |