Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CML37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_123603.3 | NP_199053.1 | 1 | 1029 | 99 | |
GenBank mRNA | AY093775.1 | - | 1 | 575 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |