Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDG25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344444.1 | NP_001318731.1 | 3 - 4 | 983 | 80 | |
NM_001344445.1 | NP_001330664.1 | 3 - 4 | 985 | 80 | ||
NM_001344446.1 | NP_001330665.1 | 3 - 4 | 974 | 80 | ||
NM_001344447.1 | NP_001330668.1 | 3 - 4 | 982 | 80 | ||
NM_001344448.1 | NP_001330669.1 | 3 - 4 | 982 | 80 | ||
NM_001344449.1 | NP_001330666.1 | 3 - 4 | 983 | 80 | ||
NM_001344450.1 | NP_001330667.1 | 2 - 3 | 805 | 80 | ||
NM_001344451.1 | NP_001330670.1 | 3 - 4 | 981 | 80 | ||
GenBank mRNA | EU014691.1 | - | 2 - 3 | 774 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |