Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_123624.4 | NP_199074.1 | 1 - 2 | 324 | 68 | |
GenBank mRNA | BT020312.1 | - | 1 - 2 | 260 | 68 | |
BT020456.1 | - | 1 - 2 | 211 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |