Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DFR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_123645.4 | NP_199094.1 | 5 - 6 | 996 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |