Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_123663.3 | NP_199111.1 | 7 - 8 | 921 | 83 | |
GenBank mRNA | AF395060.1 | - | 7 - 8 | 754 | 83 | |
AK176360.1 | - | 7 - 8 | 836 | 83 | ||
BT025548.1 | - | 7 - 8 | 754 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |