Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ETFBETA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036925.2 | NP_001032002.1 | 1 - 2 | 123 | 95 | |
NM_001344482.1 | NP_001332757.1 | 1 - 2 | 204 | 95 | ||
NM_123709.4 | NP_199156.1 | 1 - 2 | 140 | 95 | ||
GenBank mRNA | AF446867.1 | - | 1 - 2 | 65 | 95 | |
AK317065.1 | - | 1 - 2 | 106 | 95 | ||
AY045657.1 | - | 1 - 2 | 114 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |