Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UAH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_123726.4 | NP_199173.2 | 5 - 6 | 793 | 94 | |
GenBank mRNA | AK230338.1 | - | 1759 | 94 | ||
AY074343.1 | - | 5 - 6 | 757 | 94 | ||
AY091387.1 | - | 5 - 6 | 582 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |