Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TZP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344491.1 | NP_001330182.1 | 1 - 2 | 936 | 152 | |
NM_001344492.1 | NP_001330183.1 | 2 - 3 | 765 | 152 | ||
NM_001344493.1 | NP_001330181.1 | 2 - 3 | 471 | 152 | ||
NM_123729.3 | NP_199176.1 | 2 - 3 | 486 | 152 | ||
GenBank mRNA | BT003987.1 | - | 2 - 3 | 464 | 152 | |
BT005699.1 | - | 1 - 2 | 335 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |