Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344531.1 | NP_001332549.1 | 7 - 8 | 1540 | 82 | |
NM_123774.4 | NP_199220.1 | 7 - 8 | 1270 | 82 | ||
GenBank mRNA | AF093753.1 | - | 7 - 8 | 1100 | 82 | |
AF135155.1 | - | 7 - 8 | 1148 | 82 | ||
AY039951.1 | - | 7 - 8 | 1218 | 82 | ||
AY048257.1 | - | 7 - 8 | 1209 | 82 | ||
AY079384.1 | - | 7 - 8 | 1100 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |