Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RING1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161288.1 | NP_001154760.1 | 8 - 9 | 820 | 87 | |
NM_001344539.1 | NP_001331587.1 | 5 - 6 | 386 | 87 | ||
NM_123795.5 | NP_199241.2 | 6 - 7 | 860 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |