Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344547.1 | NP_001331409.1 | 7 - 8 | 1104 | 134 | |
NM_001344548.1 | NP_001331410.1 | 7 - 8 | 1023 | 134 | ||
NM_001344549.1 | NP_001331408.1 | 6 - 7 | 647 | 134 | ||
NM_123815.1 | NP_199261.1 | 5 - 6 | 719 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |