Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344589.1 | NP_001330210.1 | 6 - 7 | 936 | 77 | |
NM_123881.4 | NP_199326.1 | 6 - 7 | 830 | 77 | ||
GenBank mRNA | AF389298.1 | - | 6 - 7 | 693 | 77 | |
AY097362.1 | - | 6 - 7 | 506 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |