Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRPC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344590.1 | NP_001332060.1 | 38 - 39 | 3693 | 59 | |
NM_123882.5 | NP_199327.4 | 38 - 39 | 3611 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |