Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RTL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344592.1 | NP_001330579.1 | 5 - 6 | 1912 | 113 | |
NM_001344593.1 | NP_001330578.1 | 5 - 6 | 1837 | 113 | ||
NM_123883.2 | NP_199328.1 | 6 - 7 | 1674 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |