Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAS41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_123928.4 | NP_199373.1 | 2 - 3 | 448 | 134 | |
GenBank mRNA | AY072123.1 | - | 2 - 3 | 344 | 134 | |
AY096486.1 | - | 2 - 3 | 187 | 134 | ||
AY463617.1 | - | 2 - 3 | 187 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |