Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APG7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_123958.3 | NP_568652.1 | 7 - 8 | 1608 | 75 | |
GenBank mRNA | AB073173.1 | - | 7 - 8 | 1561 | 75 | |
AF492761.1 | - | 7 - 8 | 1479 | 75 | ||
AY034945.1 | - | 7 - 8 | 1539 | 75 | ||
AY150456.1 | - | 7 - 8 | 1479 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |