Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KASI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036941.1 | NP_001032018.1 | 4 - 5 | 1014 | 63 | |
NM_001203550.1 | NP_001190479.1 | 3 - 4 | 1186 | 63 | ||
NM_123998.3 | NP_199441.1 | 3 - 4 | 1283 | 63 | ||
GenBank mRNA | AY037261.1 | - | 3 - 4 | 1173 | 63 | |
AY087843.1 | - | 3 - 4 | 1148 | 63 | ||
AY094005.1 | - | 3 - 4 | 1073 | 63 | ||
AY123979.1 | - | 3 - 4 | 1177 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |