Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRP39-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344676.1 | NP_001330626.1 | 4 - 5 | 926 | 91 | |
NM_001344677.1 | NP_001330627.1 | 2 - 3 | 548 | 91 | ||
NM_124010.3 | NP_199452.2 | 6 - 7 | 1185 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |