Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRP39-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344676.1 | NP_001330626.1 | 7 - 8 | 1288 | 107 | |
NM_001344677.1 | NP_001330627.1 | 5 - 6 | 910 | 107 | ||
NM_124010.3 | NP_199452.2 | 9 - 10 | 1547 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |