Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124045.5 | NP_199487.1 | 2 - 3 | 349 | 111 | |
GenBank mRNA | AY099693.1 | - | 2 - 3 | 273 | 111 | |
AY128872.1 | - | 2 - 3 | 185 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |