Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124050.2 | NP_568669.1 | 3 - 4 | 430 | 103 | |
GenBank mRNA | DQ447047.1 | - | 3 - 4 | 401 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |