Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CBL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344708.1 | NP_001330804.1 | 7 - 8 | 990 | 143 | |
NM_124081.4 | NP_199521.1 | 8 - 9 | 923 | 143 | ||
GenBank mRNA | AB060590.1 | - | 7 - 8 | 577 | 143 | |
AF411958.1 | - | 7 - 8 | 577 | 143 | ||
BT002995.1 | - | 8 - 9 | 755 | 143 | ||
BT004458.1 | - | 7 - 8 | 577 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |