Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124091.5 | NP_568678.1 | 5 - 6 | 420 | 78 | |
GenBank mRNA | AF324990.2 | - | 5 - 6 | 370 | 78 | |
AK227103.1 | - | 5 - 6 | 383 | 78 | ||
AY080753.1 | - | 5 - 6 | 349 | 78 | ||
AY085384.1 | - | 5 - 6 | 374 | 78 | ||
AY114013.1 | - | 4 - 5 | 250 | 78 | ||
BT023739.1 | - | 4 - 5 | 250 | 78 | ||
BT025777.1 | - | 4 - 5 | 250 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |