Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TDT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124129.4 | NP_199567.1 | 2 - 3 | 2345 | 73 | |
GenBank mRNA | AB043024.1 | - | 2 - 3 | 1622 | 73 | |
AY072145.1 | - | 2 - 3 | 1640 | 73 | ||
AY133792.1 | - | 2 - 3 | 1589 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |