Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAUT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124152.3 | NP_568688.1 | 8 - 9 | 1689 | 79 | |
GenBank mRNA | AY056202.1 | - | 8 - 9 | 1637 | 79 | |
AY091394.1 | - | 8 - 9 | 1572 | 79 | ||
KJ138647.1 | - | 8 - 9 | 1572 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |