Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344749.1 | NP_001330051.1 | 3 - 4 | 623 | 79 | |
NM_124158.4 | NP_199595.1 | 3 - 4 | 694 | 79 | ||
GenBank mRNA | AY049305.1 | - | 3 - 4 | 565 | 79 | |
BT001036.1 | - | 3 - 4 | 523 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |