Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THAS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085264.2 | NP_001078733.1 | 2 - 3 | 553 | 73 | |
NM_124175.4 | NP_199612.3 | 2 - 3 | 430 | 73 | ||
GenBank mRNA | AY327541.1 | - | 2 - 3 | 389 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |