Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161296.1 | NP_001154768.1 | 6 - 7 | 1058 | 66 | |
NM_001344757.1 | NP_001318757.1 | 7 - 8 | 1099 | 66 | ||
NM_124183.4 | NP_199620.4 | 6 - 7 | 924 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |