Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124205.4 | NP_199641.1 | 4 - 5 | 1206 | 124 | |
GenBank mRNA | AY049265.1 | - | 4 - 5 | 1182 | 124 | |
AY065428.1 | - | 4 - 5 | 1148 | 124 | ||
AY090283.1 | - | 4 - 5 | 1118 | 124 | ||
AY096379.1 | - | 4 - 5 | 1118 | 124 | ||
U72351.1 | - | 4 - 5 | 1153 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |