Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sks3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344773.1 | NP_001332438.1 | 4 - 5 | 715 | 66 | |
NM_124220.3 | NP_199656.2 | 4 - 5 | 700 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |