Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203563.2 | NP_001190492.1 | 24 - 25 | 3498 | 95 | |
NM_124236.3 | NP_199671.1 | 24 - 25 | 3340 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |