Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124278.4 | NP_199712.2 | 4 - 5 | 720 | 101 | |
GenBank mRNA | AK221044.1 | - | 4 - 5 | 720 | 101 | |
BT010406.1 | - | 4 - 5 | 487 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |