Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OVA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036965.2 | NP_001032042.1 | 9 - 10 | 1218 | 73 | |
NM_001203568.1 | NP_001190497.1 | 10 - 11 | 1651 | 73 | ||
NM_124280.4 | NP_199714.2 | 9 - 10 | 1330 | 73 | ||
GenBank mRNA | AY080830.1 | - | 9 - 10 | 1147 | 73 | |
AY142605.1 | - | 9 - 10 | 1047 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |