Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IMPA-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124308.2 | NP_199742.1 | 1 - 2 | 367 | 66 | |
GenBank mRNA | DQ447054.1 | - | 1 - 2 | 211 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |