Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BXL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124313.3 | NP_199747.1 | 4 - 5 | 1169 | 48 | |
GenBank mRNA | AY120767.1 | - | 4 - 5 | 1046 | 48 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |