Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bZIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124322.3 | NP_199756.1 | 1 | 741 | 86 | |
GenBank mRNA | AY088207.1 | - | 1 | 734 | 86 | |
AY136307.1 | - | 1 | 741 | 86 | ||
BT000400.1 | - | 1 | 425 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |