Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AAP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124341.4 | NP_199774.1 | 2 - 3 | 589 | 109 | |
GenBank mRNA | AK229102.1 | - | 2 - 3 | 345 | 109 | |
X95736.1 | - | 385 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Arabidopsis thaliana Probe spans exons |