Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GH9A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124350.3 | NP_199783.1 | 1 - 2 | 675 | 102 | |
GenBank mRNA | AF074375.1 | - | 1 - 2 | 438 | 102 | |
U37702.1 | - | 1 - 2 | 445 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |