Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124455.2 | NP_199890.1 | 2 - 3 | 398 | 66 | |
GenBank mRNA | BT020525.1 | - | 2 - 3 | 390 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |