Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124455.2 | NP_199890.1 | 3 - 4 | 493 | 108 | |
GenBank mRNA | BT020525.1 | - | 3 - 4 | 485 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |