Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124455.2 | NP_199890.1 | 4 - 5 | 620 | 64 | |
GenBank mRNA | BT020525.1 | - | 4 - 5 | 612 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |