Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124463.4 | NP_199898.1 | 14 - 15 | 1330 | 68 | |
GenBank mRNA | AY070728.1 | - | 14 - 15 | 1165 | 68 | |
BT000839.1 | - | 13 - 14 | 1037 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |