Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124463.4 | NP_199898.1 | 7 - 8 | 655 | 77 | |
GenBank mRNA | AY070728.1 | - | 7 - 8 | 490 | 77 | |
BT000839.1 | - | 6 - 7 | 362 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |