Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLPC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124471.4 | NP_568746.1 | 2 - 3 | 1044 | 68 | |
GenBank mRNA | AK227173.1 | - | 2 - 3 | 737 | 68 | |
AY102125.1 | - | 2 - 3 | 709 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |