Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PABN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124491.3 | NP_568751.1 | 2 - 3 | 479 | 63 | |
GenBank mRNA | AY054186.1 | - | 2 - 3 | 320 | 63 | |
AY057580.1 | - | 2 - 3 | 307 | 63 | ||
AY079392.1 | - | 2 - 3 | 261 | 63 | ||
AY116673.1 | - | 2 - 3 | 304 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |