Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SWI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344934.1 | NP_001332209.1 | 5 - 6 | 1423 | 141 | |
NM_124512.3 | NP_568757.2 | 6 - 7 | 1425 | 141 | ||
GenBank mRNA | AF362001.1 | - | 5 - 6 | 1382 | 141 | |
AF466153.1 | - | 6 - 7 | 1424 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |