Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EYE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124522.4 | NP_199956.1 | 8 - 9 | 2112 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |