Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KEA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125943.1 | NP_001119415.1 | 18 - 19 | 1703 | 89 | |
NM_124550.2 | NP_568763.1 | 18 - 19 | 1834 | 89 | ||
GenBank mRNA | AY070064.1 | - | 18 - 19 | 1649 | 89 | |
AY084820.1 | - | 18 - 19 | 1702 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |