Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KEA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125943.1 | NP_001119415.1 | 8 - 9 | 824 | 70 | |
NM_124550.2 | NP_568763.1 | 8 - 9 | 955 | 70 | ||
GenBank mRNA | AY070064.1 | - | 8 - 9 | 770 | 70 | |
AY084820.1 | - | 8 - 9 | 823 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |